Learn More
Lauda™ Metal Hoses
Brand:  Lauda™ LZM 092
Additional Details : Weight : 3.50000kg
Product Code. 15205537
  • 1060.00€ / Each
Estimated Shipment: 05-06-2024
to see stock.
Specifications
-100°C to +350°C
Metal Hose
200 cm