Learn More
Brady™ Metalized MiniMark™ Polyester Name and Rating Plates Labels
Product Code. 11721825
Change view
Click to view available options
Width (Metric):
25.4 mm
38.1 mm
50.8 mm
69.85 mm
76.2 mm
Height (Metric):
12.70 mm
19.05 mm
25.40 mm
31.75 mm
50.00 mm
Unit Size:
Each
Product Code. Width (Metric) Height (Metric) unitSize
11721825 25.4 mm 12.70 mm Each
11751825 38.1 mm 19.05 mm Each
11781785 69.85 mm 31.75 mm Each
11761795 50.8 mm 25.40 mm Each
11741735 76.2 mm 50.00 mm Each
5 options
Product Code. 11721825
Brand: Brady™ 710081
Identify your Asset and Rating Plates with these metallic looking MiniMark™ Polyester Name and Rating Plates Labels.
  • Identify your Asset and Rating Plates with these metallic looking labels.
Specifications
Certifications/Compliance RoHS, 2011/65/EU
Description MiniMark Name and Rating Plates Labels
Adhesive Type Acrylic, Permanent
Coating None
Color Silver
Display Blank
Finish Satin
Format Die-cut
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset tracking, General Identification
For Use With (Equipment) MiniMark Printer
Horizontal Repeat (Metric) 28.4 mm
Labels per Row 3
Min. Operating Temperature (Metric) -40°C
No. per Pack 2865 Labels/Box
Quantity 2865 Pk.
Shape Rectangle
Size (Metric) 25.4 x 12.7 mm (W x H)
Temperature Range (English) -40°F to +275°F
Temperature Range (Metric) -40°C to +135°C
Thickness (English) 0.003 in.
Thickness (Metric) 0.075 mm
Vertical Repeat (Metric) 15.70 mm
Dimensions (LxW) 12.7 x 25.4 mm
Length (English) 0.5 in.
Length (Metric) 12.7 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (Metric) 12.70 mm
Alternate Part Number MNK-BPT-5-7563