missing translation for 'onlineSavingsMsg'
Learn More

Metamizol Sodium Monohydrate, TRC

Product Code. 17950365
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17950365 500 mg 500mg
17944065 250 mg 250mg
17940375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17950365 Supplier Toronto Research Chemicals Supplier No. TRCM244740500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5907-38-0
InChI Formula InChI=1S/C13H17N3O4S.Na.H2O/c1-10-12(14(2)9-21(18,19)20)13(17)16(15(10)3)11-7-5-4-6-8-11;;/h4-8H,9H2,1-3H3,(H,18,19,20);;1H2/q;+1;/p-1
IUPAC Name sodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-methylamino]methanesulfonate;hydrate
Molecular Formula C13H16N3O4S.Na.H2O
Molecular Weight (g/mol) 351.35
Chemical Name or Material Metamizole Sodium Monohydrate

Specifications

Quantity 500 mg
Synonym Metamizole Sodium Monohydrate, 1-[(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-methanesulfonic acid monosodium salt monohydrate, (Antipyrinylmethylamino)-methanesulfonic acid sodium salt monohydrate, [(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-methanesulfonic acid sodium salt monohydrate, Dipyrone monohydrate, Metamizol monohydrate, Sodium (antipyrinylmethylamino)methanesulfonate hydrate
SMILES O.[Na+].CN(CS(=O)(=O)[O-])C1=C(C)N(C)N(C1=O)c2ccccc2
Formula Weight 351.0865
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.