missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Metamizole sodium monohydrate

Product Code. 17291196 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17291196 250 mg 250mg
17281196 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 17291196 Supplier Thermo Scientific Chemicals Supplier No. 467482500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5907-38-0
Molecular Formula C13H18N3NaO5S

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: soluble. Other solubilities: soluble in methanol, insoluble in ether, acetone, chloroform
Formula Weight 351.36
InChI Key UNZIDPIPYUMVPA-UHFFFAOYSA-M
SMILES O.[Na+].CN(CS([O-])(=O)=O)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1
IUPAC Name sodium [(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methyl)amino]methanesulfonate hydrate
Molecular Weight (g/mol) 351.35
Chemical Name or Material Metamizole sodium monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.