Learn More
Methanol, LC/MS grade, Thermo Scientific
36.30€ - 115.00€
Products 3
Product Code Brand dimQuantity Price Quantity & Availability  
Product Code Brand dimQuantity Price Quantity & Availability  
30280930
View Documents Promotion Details
Thermo Scientific
T004560500
ONLINE EXCLUSIVE PRICE
38.50€
36.30€ / 500mL
Save 2.20€
5% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 Restricted Item
30280931
View Documents Promotion Details
Thermo Scientific
T004561000
ONLINE EXCLUSIVE PRICE
75.20€
55.10€ / 1 L
Save 20.10€
26% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 Restricted Item
30280932
View Documents Promotion Details
Thermo Scientific
T004562500
ONLINE EXCLUSIVE PRICE
163.00€
115.00€ / 2.5 L
Save 48.00€
29% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 Restricted Item
Description
Description
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Methanol, LC/MS grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Methanol