missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methsuximide, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Methsuximide
Specifications
Specifications
| Chemical Name or Material | Methsuximide |
| CAS | 77-41-8 |
| Analyte or Component Names | Methsuximide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H13 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2,5-Pyrrolidinedione, 1,3-dimethyl-3-phenyl-, Succinimide, N,2-dimethyl-2-phenyl- (6CI,7CI,8CI), 1,3-Dimethyl-3-phenylsuccinimide, Celontin, Mesuximide, Mesuximidum, Methsuximid, Methsuximide, N,2-Dimethyl-2-phenylsuccinimide, PM 396, Petinutin, Racemic methsuximide, (+/-)-Mesuximide |
| InChI Formula | InChI=1S/C12H13NO2/c1-12(9-6-4-3-5-7-9)8-10(14)13(2)11(12)15/h3-7H,8H2,1-2H3 |
| SMILES | CN1C(=O)CC(C)(C1=O)c2ccccc2 |
| IUPAC Name | 1,3-dimethyl-3-phenylpyrrolidine-2,5-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction