missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-chloro-3-iodobenzoate, 98+%

Product Code. 11359016
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359016 5 g 5g
11369016 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359016 Supplier Thermo Scientific Alfa Aesar Supplier No. L20188.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 289039-84-5 | C8H7ClINO2 | 311.503 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 289039-84-5
Molecular Formula C8H7ClINO2
Molecular Weight (g/mol) 311.503
MDL Number MFCD00673017
InChI Key VTDNWMZYPVQPCN-UHFFFAOYSA-N
PubChem CID 7020924
IUPAC Name methyl 2-amino-5-chloro-3-iodobenzoate
SMILES COC(=O)C1=CC(=CC(=C1N)I)Cl

Specifications

Melting Point 67°C to 69°C
Quantity 5 g
Sensitivity Light sensitive
Synonym 2-amino-5-chloro-3-iodobenzoic acid methyl ester, pubchem3349, acmc-1cp8o, methyl 3-iodo-5-chloroanthranilate, 4-chloro-2-iodo-6-methoxycarbonyl aniline, methyl 2-amino-5-chloro-3-iodobenzoate, methyl 2-azanyl-5-chloranyl-3-iodanyl-benzoate, 2-amino-5-chloro-3-iodo-benzoic acid methyl ester, benzoic acid,2-amino-5-chloro-3-iodo-, methyl ester, benzoic acid, 2-amino-5-chloro-3-iodo-, methyl ester
Formula Weight 311.51
Percent Purity ≥98%
Chemical Name or Material Methyl 2-amino-5-chloro-3-iodobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.