missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Specifications
Melting Point | 85°C to 88°C |
Flash Point | >100°C |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 25 g |
Solubility Information | Solubility in water: .. Other solubilities: soluble in methanol |
Formula Weight | 171.22 |
Percent Purity | 98% |
Chemical Name or Material | Methyl 3-amino-4-methyl-2-thiophenecarboxylate |
Safety and Handling
Product Identifier
- Methyl 3-amino-4-methylthiophene-2-carboxylate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'