missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chlorobenzoate, 98%, Thermo Scientific Chemicals

Catalog Number p-7025726
51.60 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2905-65-9
C8H7ClO2
170.59
MFCD00000592
XRDRKVPNHIWTBX-UHFFFAOYSA-N
methyl m-chlorobenzoate, benzoic acid, 3-chloro-, methyl ester, m-chlorobenzoic acid methyl ester, methyl-3-chlorobenzoate, benzoic acid, m-chloro-, methyl ester, 3-chlorobenzoic acid methyl ester, 3-chloro-benzoic acid methyl ester, acmc-1cegg, dsstox_cid_7507, 3-chlorobenzoic acid methyl
17946
methyl 3-chlorobenzoate
COC(=O)C1=CC=CC(Cl)=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2905-65-9
C8H7ClO2
170.59
MFCD00000592
XRDRKVPNHIWTBX-UHFFFAOYSA-N
methyl m-chlorobenzoate, benzoic acid, 3-chloro-, methyl ester, m-chlorobenzoic acid methyl ester, methyl-3-chlorobenzoate, benzoic acid, m-chloro-, methyl ester, 3-chlorobenzoic acid methyl ester, 3-chloro-benzoic acid methyl ester, acmc-1cegg, dsstox_cid_7507, 3-chlorobenzoic acid methyl
17946
methyl 3-chlorobenzoate
COC(=O)C1=CC=CC(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 20°C to 22°C
Density 1.227
Boiling Point 99°C to 101°C (12 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.531
Quantity 25 g
Beilstein 2045126
Merck Index 14,2123
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material Methyl 3-chlorobenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'