missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyclohexenecarboxylate, 97%, Thermo Scientific™

Catalog Number 15269786
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
6493-77-2
C8H12O2
140.18
MFCD00053266
IPUNVLFESXFVFH-UHFFFAOYNA-N
methyl cyclohex-3-enecarboxylate, 3-cyclohexene-1-carboxylic acid methyl ester, methyl 3-cyclohexenecarboxylate, 3-cyclohexene-1-carboxylic acid, methyl ester, methyl 3-cyclohexene-1-carboxylate, 1-carbomethoxy-3-cyclohexene, methyl 4-cyclohexenecarboxylate, methyl 1,2,3,6-tetrahydrobenzoate, 1,3-butadiene-methyl acrylate adduct, acmc-1bity
96926
methyl cyclohex-3-ene-1-carboxylate
COC(=O)C1CCC=CC1
This item is not returnable. View return policy
6493-77-2
C8H12O2
140.18
MFCD00053266
IPUNVLFESXFVFH-UHFFFAOYNA-N
methyl cyclohex-3-enecarboxylate, 3-cyclohexene-1-carboxylic acid methyl ester, methyl 3-cyclohexenecarboxylate, 3-cyclohexene-1-carboxylic acid, methyl ester, methyl 3-cyclohexene-1-carboxylate, 1-carbomethoxy-3-cyclohexene, methyl 4-cyclohexenecarboxylate, methyl 1,2,3,6-tetrahydrobenzoate, 1,3-butadiene-methyl acrylate adduct, acmc-1bity
96926
methyl cyclohex-3-ene-1-carboxylate
COC(=O)C1CCC=CC1
Density 1.0200g/mL
Boiling Point 185.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 500 mL
Specific Gravity 1.02
Formula Weight 140.18
Percent Purity 97%
Chemical Name or Material Methyl 3-cyclohexenecarboxylate
compliance-icons
Product Identifier
  • Methyl 3-cyclohexenecarboxylate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'