missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-2-methoxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11342949
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11342949

Brand: Thermo Scientific Alfa Aesar H27773.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139102-34-4
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD06797867
InChI Key WPGAGRPPDYAZAD-UHFFFAOYSA-N
PubChem CID 15128242
IUPAC Name methyl 4-bromo-2-methoxybenzoate
SMILES COC1=C(C=CC(=C1)Br)C(=O)OC

Specifications

Melting Point 32°C to 34°C
Boiling Point 100°C to 108°C (0.1 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Synonym methyl4-bromo-2-methoxybenzoate, benzoic acid, 4-bromo-2-methoxy-, methyl ester, 4-bromo-2-methoxybenzoic acid methyl ester, ablock ab-14-0789, benzoic acid,4-bromo-2-methoxy-, methyl ester, pubchem16069, acmc-209ciq, methyl-4-bromo-2-methoxybenzoate, zerenex e/1100188, 4-bromo-2-methoxybenzoate 98
Formula Weight 245.08
Percent Purity 98%
Chemical Name or Material Methyl 4-bromo-2-methoxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.