missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(trifluoromethyl)benzoate, 98%

Product Code. 11428366
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11428366

Brand: Thermo Scientific Alfa Aesar A15400.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2967-66-0 | C9H7F3O2 | 204.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2967-66-0
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00042324
InChI Key VAZWXPJOOFSNLB-UHFFFAOYSA-N
Synonym methyl 4-trifluoromethyl benzoate, methyl-4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid methyl ester, methyl-4-trifluoromethylbenzoate, 3-cf3-c6h4-cooch3, methyl p-trifluoromethyl benzoate, methyl alpha,alpha,alpha-trifluoro-p-toluate, benzoic acid, 4-trifluoromethyl-, methyl ester, pubchem14120, acmc-209hab
PubChem CID 520447
IUPAC Name methyl 4-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 13°C to 14°C
Density 1.268
Boiling Point 86°C to 87°C (10 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.452
Quantity 5 g
Beilstein 1963288
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material Methyl 4-(trifluoromethyl)benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.