Learn More
Methyl 4-(trifluoromethyl)benzoate, 98%
Product Code. 11448366
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11448366
Brand: Thermo Scientific Alfa Aesar A15400.22
Request Bulk or Custom Format
CAS: 2967-66-0 | C9H7F3O2 | 204.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2967-66-0
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00042324
InChI Key VAZWXPJOOFSNLB-UHFFFAOYSA-N
Synonym methyl 4-trifluoromethyl benzoate, methyl-4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid methyl ester, methyl-4-trifluoromethylbenzoate, 3-cf3-c6h4-cooch3, methyl p-trifluoromethyl benzoate, methyl alpha,alpha,alpha-trifluoro-p-toluate, benzoic acid, 4-trifluoromethyl-, methyl ester, pubchem14120, acmc-209hab
PubChem CID 520447
IUPAC Name methyl 4-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C(F)(F)F
Specifications
Melting Point 13°C to 14°C
Density 1.268
Boiling Point 86°C to 87°C (10 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.452
Quantity 100 g
Beilstein 1963288
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material Methyl 4-(trifluoromethyl)benzoate