missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-7055758
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
50593-91-4
C7H7BrN2O2S
263.11
MFCD05664428
MYZJIEWTRJTWCD-UHFFFAOYSA-N
methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate, methyl 5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylate, methyl 5-bromo-2-methylthio-4-pyrimidinecarboxylate, methylbromomethylsulfanylpyrimidinecarboxylate, methyl 5-bromo-2-methylthiopyrimidine-4-carboxylate, methyl-5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarb, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate,97, 5-bromo-2-methylsulfanylpyrimidine-4-carboxylic acid methyl ester
2794729
COC(=O)C1=NC(SC)=NC=C1Br
This item is not returnable. View return policy
50593-91-4
C7H7BrN2O2S
263.11
MFCD05664428
MYZJIEWTRJTWCD-UHFFFAOYSA-N
methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate, methyl 5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylate, methyl 5-bromo-2-methylthio-4-pyrimidinecarboxylate, methylbromomethylsulfanylpyrimidinecarboxylate, methyl 5-bromo-2-methylthiopyrimidine-4-carboxylate, methyl-5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarb, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate,97, 5-bromo-2-methylsulfanylpyrimidine-4-carboxylic acid methyl ester
2794729
COC(=O)C1=NC(SC)=NC=C1Br

It can be used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water (0.42 g/L).

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 61°C to 64°C
Color Yellow
Quantity 250 mg
UN Number UN2811
Beilstein 910703
Solubility Information Slightly soluble in water (0.42g/L).
IUPAC Name methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate
Formula Weight 263.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'