missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals

Catalog Number 15407868
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
89581-38-4
C6H5BrN2O2
217.02
MFCD11111606
XILAKTMDKMVJQV-UHFFFAOYSA-N
methyl 5-bromopyrimidine-2-carboxyate, methyl-5-bromo-2 pyrimidine carboxylate, 5-bromo-pyrimidine-2-carboxylic acid methyl ester, 2-pyrimidinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrimidinecarboxylicacid, 5-bromo-, methyl ester, 5-bromopyrimidine-2-carboxylic acid methyl ester, acmc-20dmbr, pubchem21032, methyl5-bromopyrimidine-2-carboxylate, methyl 5-bromanylpyrimidine-2-carboxylate
45790831
methyl 5-bromopyrimidine-2-carboxylate
COC(=O)C1=NC=C(Br)C=N1
This item is not returnable. View return policy
89581-38-4
C6H5BrN2O2
217.02
MFCD11111606
XILAKTMDKMVJQV-UHFFFAOYSA-N
methyl 5-bromopyrimidine-2-carboxyate, methyl-5-bromo-2 pyrimidine carboxylate, 5-bromo-pyrimidine-2-carboxylic acid methyl ester, 2-pyrimidinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrimidinecarboxylicacid, 5-bromo-, methyl ester, 5-bromopyrimidine-2-carboxylic acid methyl ester, acmc-20dmbr, pubchem21032, methyl5-bromopyrimidine-2-carboxylate, methyl 5-bromanylpyrimidine-2-carboxylate
45790831
methyl 5-bromopyrimidine-2-carboxylate
COC(=O)C1=NC=C(Br)C=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 217.02
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 5-bromopyrimidine-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'