Learn More
Methyl 5-chloro-2-nitrobenzoate, 98%, Thermo Scientific™
Product Code. 11469396
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11469396
Brand: Thermo Scientific Alfa Aesar A19098.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51282-49-6
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.589
MDL Number MFCD00272139
InChI Key JGBJHRKCUKTQOE-UHFFFAOYSA-N
Synonym 5-chloro-2-nitrobenzoic acid methyl ester, benzoic acid, 5-chloro-2-nitro-, methyl ester, unii-1t5d4k1v51, methyl 5-chloro-2-nitro-benzoate, pubchem3712, acmc-209ksr, dsstox_cid_24948, dsstox_rid_80608, dsstox_gsid_44948, ksc495i7r
PubChem CID 171018
IUPAC Name methyl 5-chloro-2-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Cl)[N+](=O)[O-]
Specifications
Melting Point 48°C to 50°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 215.59
Percent Purity 98%
Chemical Name or Material Methyl 5-chloro-2-nitrobenzoate