missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-bromophenylacetate, 97%, Thermo Scientific™

Catalog Number p-7058329
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3042-81-7
C9H9BrO2
229.073
MFCD00013535
NHFBYYMNJUMVOT-UHFFFAOYSA-N
methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate
137806
methyl 2-bromo-2-phenylacetate
COC(=O)C(C1=CC=CC=C1)Br
This item is not returnable. View return policy
3042-81-7
C9H9BrO2
229.073
MFCD00013535
NHFBYYMNJUMVOT-UHFFFAOYSA-N
methyl alpha-bromophenylacetate, methyl bromophenylacetate, methyl bromo phenyl acetate, methyl .alpha.-bromophenylacetate, methyl2-bromo-2-phenylacetate, methyl .alpha.-bromobenzeneacetate, benzeneacetic acid, .alpha.-bromo-, methyl ester, +/--methyl alpha-bromophenylacetate, alpha-bromophenylacetic acid methyl ester, methyl .alpha.-bromo-.alpha.-phenylacetate
137806
methyl 2-bromo-2-phenylacetate
COC(=O)C(C1=CC=CC=C1)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 90°C to 92°C (0.5 mmHg)
Flash Point 150°C (302°F)
Quantity 5 g
UN Number UN3265
Beilstein 1102743
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material Methyl alpha-bromophenylacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'