Learn More
Thermo Scientific Chemicals Methyl cellulose, viscosity 4000 cPs
Description
Methyl cellulose is used as a thickener and an emulsifier in various food and cosmetic products; as a gel in advanced cookery and as a lubricant. It is also involved in the treatment of constipation. It acts as a buffer additive in capillary electrophoresis to control electoosmotic flow for improved separations. In paper and textile industries, it is used as a sizing, thereby protecting the fibers from water or oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Odor | Odorless |
Quantity | 500 g |
Merck Index | 14,6040 |
Solubility Information | Soluble in cold water. |
Physical Form | Powder |
Chemical Name or Material | Methyl cellulose, Viscosity: 4000cPs |
RUO – Research Use Only