missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Green, zinc chloride

Catalog Number p-7042154
70.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7114-03-6
C27H35Cl4N3Zn
608.78
MFCD00151094
ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
C.I. 42590
131675247
zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7114-03-6
C27H35Cl4N3Zn
608.78
MFCD00151094
ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
C.I. 42590
131675247
zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]

Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Green is a DNA stain used as a tracking dye in acid buffers for electrophoresis. It helps differentiate bacteria and is also a tissue stain for nucleoli and mitochondria in tissues. It is used with Bismarck Brown frequently.

Solubility
Soluble in water, ethanol or methanol

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Beilstein 3914106
Solubility Information Soluble in water,ethanol or methanol
Formula Weight 653.24
Color Green
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl Green, zinc chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'