missing translation for 'onlineSavingsMsg'
Learn More

Methyl imidazo[1,2-a]pyridine-7-carboxylate, 95%, Thermo Scientific Chemicals

Catalog Number p-7032893
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
86718-01-6
C9H8N2O2
176.18
MFCD09910350
KYHRVKMXYXBEQN-UHFFFAOYSA-N
methyl imidazo 1,2-a pyridine-7-carboxylate, imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, 7-methoxycarbonyl imidazo 1,2-a pyridine, methylimidazo 1,2-a pyridine-7-carboxylate, methyl h-imidazo 1,2-a pyridine-7-carboxylate, imidazo 1,2-a pyridine-7-carboxylic acid, methyl ester, pubchem21685, methylimidazoapyridinecarboxylate, methyl imidazo-1,2-a pyridine-7-carboxylate, 7-imidazo 1,2-a pyridinecarboxylic acid methyl ester
15096964
methyl imidazo[1,2-a]pyridine-7-carboxylate
COC(=O)C1=CC2=NC=CN2C=C1
This item is not returnable. View return policy
86718-01-6
C9H8N2O2
176.18
MFCD09910350
KYHRVKMXYXBEQN-UHFFFAOYSA-N
methyl imidazo 1,2-a pyridine-7-carboxylate, imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, 7-methoxycarbonyl imidazo 1,2-a pyridine, methylimidazo 1,2-a pyridine-7-carboxylate, methyl h-imidazo 1,2-a pyridine-7-carboxylate, imidazo 1,2-a pyridine-7-carboxylic acid, methyl ester, pubchem21685, methylimidazoapyridinecarboxylate, methyl imidazo-1,2-a pyridine-7-carboxylate, 7-imidazo 1,2-a pyridinecarboxylic acid methyl ester
15096964
methyl imidazo[1,2-a]pyridine-7-carboxylate
COC(=O)C1=CC2=NC=CN2C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 133°C to 135°C
Quantity 250 mg
Formula Weight 176.18
Percent Purity 95%
Chemical Name or Material Methyl imidazo[1,2-a]pyridine-7-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'