Learn More
Alfa Aesar™ Methyl imidazole-4-carboxylate, 98%
Brand: Alfa Aesar™ H55932.06
Additional Details : CAS : 17325-26-7 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
17325-26-7 | |
126.115 | |
DVLGIQNHKLWSRU-UHFFFAOYSA-N | |
565661 | |
COC(=O)C1=CN=CN1 |
C5H6N2O2 | |
MFCD00216584 | |
methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863 | |
methyl 1H-imidazole-5-carboxylate |
Specifications
Methyl imidazole-4-carboxylate | |
5g | |
C5H6N2O2 | |
methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863 | |
COC(=O)C1=CN=CN1 | |
126.115 | |
126.11 |
154°C to 156°C | |
17325-26-7 | |
MFCD00216584 | |
DVLGIQNHKLWSRU-UHFFFAOYSA-N | |
methyl 1H-imidazole-5-carboxylate | |
565661 | |
98% |