Learn More
Methyl nonanoate, 95%, Thermo Scientific Chemicals
Catalog Number p-3586452
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
1731-84-6
C10H20O2
172.27
MFCD00009569
IJXHLVMUNBOGRR-UHFFFAOYSA-N
methyl pelargonate, pelargonic acid methyl ester, methyl nonylate, nonanoic acid, methyl ester, methyl n-nonanoate, nonanoic acid methyl ester, unii-7xkd6qoh68, fema no. 2724, methyl ester nonanoic acid, 7xkd6qoh68
15606
CHEBI:44499
methyl nonanoate
CCCCCCCCC(=O)OC
1731-84-6
C10H20O2
172.27
MFCD00009569
IJXHLVMUNBOGRR-UHFFFAOYSA-N
methyl pelargonate, pelargonic acid methyl ester, methyl nonylate, nonanoic acid, methyl ester, methyl n-nonanoate, nonanoic acid methyl ester, unii-7xkd6qoh68, fema no. 2724, methyl ester nonanoic acid, 7xkd6qoh68
15606
CHEBI:44499
methyl nonanoate
CCCCCCCCC(=O)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -30°C
Color Colorless
Density 0.8740g/mL
Boiling Point 213°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7C(O)OCH3
Refractive Index 1.4210 to 1.423
Quantity 5 g
Specific Gravity 0.874
Solubility Information Solubility in water: practically insoluble
Formula Weight 172.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl nonanoate