missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenylacetate, 99+%

Product Code. p-7214332 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
250 mL
1 L
Unit Size:
1 L
250mL
5mL
This item is not returnable. View return policy

Product Code. 10706361

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-41-7 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-41-7
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
InChI Key CRZQGDNQQAALAY-UHFFFAOYSA-N
IUPAC Name methyl 2-phenylacetate
SMILES COC(=O)CC1=CC=CC=C1

Specifications

Density 1.0670 g/mL
Boiling Point 220.0°C
Flash Point 95°C
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H5CH2CO2CH3
Refractive Index 1.5060 to 1.5080
Quantity 5 mL
Beilstein 09, 434
Merck Index 15, 7380
Specific Gravity 1.067
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohol
Formula Weight 150.18
Chemical Name or Material Methyl phenylacetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.