missing translation for 'onlineSavingsMsg'
Learn More

Methyl propionylacetate, 99%, Thermo Scientific Chemicals

Catalog Number p-7027818
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
30414-53-0
C6H10O3
130.14
MFCD00011705
XJMIXEAZMCTAGH-UHFFFAOYSA-N
methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester
121699
methyl 3-oxopentanoate
CCC(=O)CC(=O)OC
This item is not returnable. View return policy
30414-53-0
C6H10O3
130.14
MFCD00011705
XJMIXEAZMCTAGH-UHFFFAOYSA-N
methyl 3-oxovalerate, methyl propionylacetate, methyl 3-oxo-pentanoate, pentanoic acid, 3-oxo-, methyl ester, methyl-3-oxo pentanoate, methyl 3-oxo-n-valerate, valeric acid, 3-oxo-, methyl ester, 3-oxovaleric acid methyl ester, 3-oxo-pentanoic acid methyl ester, 3-keto-n-valeric acid methyl ester
121699
methyl 3-oxopentanoate
CCC(=O)CC(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.037
Boiling Point 73°C to 74°C (5 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.422
Quantity 25 g
Beilstein 1753684
Formula Weight 130.14
Percent Purity 99%
Chemical Name or Material Methyl propionylacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'