missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number 15495758
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1193-62-0
C6H7NO2
125.127
MFCD00817048
VONGYFFEWFJHNP-UHFFFAOYSA-N
methyl pyrrole-2-carboxylate, methyl 2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, methyl ester, unii-3beg2ck3lu, pyrrole-2-carboxylic acid methyl ester, methyl1h-pyrrole-2-carboxylate, 3beg2ck3lu, 1h-pyrrole-2-carboxylic acid,methyl ester, pubchem9141, acmc-1bsa1
136930
methyl 1H-pyrrole-2-carboxylate
COC(=O)C1=CC=CN1
This item is not returnable. View return policy
1193-62-0
C6H7NO2
125.127
MFCD00817048
VONGYFFEWFJHNP-UHFFFAOYSA-N
methyl pyrrole-2-carboxylate, methyl 2-pyrrolecarboxylate, 1h-pyrrole-2-carboxylic acid, methyl ester, unii-3beg2ck3lu, pyrrole-2-carboxylic acid methyl ester, methyl1h-pyrrole-2-carboxylate, 3beg2ck3lu, 1h-pyrrole-2-carboxylic acid,methyl ester, pubchem9141, acmc-1bsa1
136930
methyl 1H-pyrrole-2-carboxylate
COC(=O)C1=CC=CN1

It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine and as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. 

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
TRUSTED_SUSTAINABILITY
Melting Point 74°C to 78°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material Methyl pyrrole-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'