missing translation for 'onlineSavingsMsg'
Learn More

Methyl Salicylate, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10060260
Click to view available options
Quantity:
500 mL
2.5 L
This item is not returnable. View return policy
119-36-8
C8H8O3
152.149
2214
OSWPMRLSEDHDFF-UHFFFAOYSA-N
methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester
4133
CHEBI:31832
methyl 2-hydroxybenzoate
COC(=O)C1=CC=CC=C1O
This item is not returnable. View return policy
119-36-8
C8H8O3
152.149
2214
OSWPMRLSEDHDFF-UHFFFAOYSA-N
methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester
4133
CHEBI:31832
methyl 2-hydroxybenzoate
COC(=O)C1=CC=CC=C1O

Methyl Salicylate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Melting Point -8°C
Color Yellow
Boiling Point 222°C
Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Formula Weight 152.15g/mol
Vapor Pressure 0.118mmHg 25
Physical Form Liquid
Product Identifier
  • Methyl salicylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'