missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-(trifluoromethyl)nicotinate, 97%, Thermo Scientific™

Catalog Number 10597970
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
221313-10-6
C8H6F3NO2
205.13
MFCD03001346
NEATXKNYTRXHQL-UHFFFAOYSA-N
2775647
methyl 6-(trifluoromethyl)pyridine-3-carboxylate
COC(=O)C1=CN=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
221313-10-6
C8H6F3NO2
205.13
MFCD03001346
NEATXKNYTRXHQL-UHFFFAOYSA-N
2775647
methyl 6-(trifluoromethyl)pyridine-3-carboxylate
COC(=O)C1=CN=C(C=C1)C(F)(F)F
Melting Point 58°C to 60.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym methyl 6-trifluoromethyl nicotinate, methyl 6-trifluoromethyl pyridine-3-carboxylate, methyl6-trifluoromethyl nicotinate, 3-pyridinecarboxylic acid, 6-trifluoromethyl-, methyl ester, methyl 6-trifluoromethylnicotinate, methyl 2-trifluoromethyl pyridine-5-carboxylate, methyl 2-trifluoromethyl-5-pyridinecarboxylate, 6-trifluoromethylnicotinic acid methyl ester, pubchem18762, methyltrifluoromethylnicotinate
Formula Weight 205.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyl 6-(trifluoromethyl)nicotinate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 6-(trifluoromethyl)nicotinate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'