missing translation for 'onlineSavingsMsg'
Learn More

Methylthymol Blue Sodium Salt, Honeywell Fluka™

Catalog Number p-7120586
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
1945-77-3
C37H44N2Na4O13S
848.779
MFCD00148905
LGVVZVZPOQJZKT-UHFFFAOYSA-N
131850582
2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]
This item is not returnable. View return policy
1945-77-3
C37H44N2Na4O13S
848.779
MFCD00148905
LGVVZVZPOQJZKT-UHFFFAOYSA-N
131850582
2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Linear Formula C37H40N2O13SNa4
UN Number NONH for all modes of transport
Formula Weight 844.74g/mol
Packaging Glass Bottle
Quantity 1 g
Chemical Name or Material Methylthymol Blue Sodium Salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'