missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylthymol Blue Sodium Salt (Technical Grade), TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Methylthymol Blue Sodium Salt |
| CAS | 1945-77-3 |
| Molecular Formula | C37H40N2Na4O13S |
| Quantity | 10 g |
| Synonym | 3H-2,1-Benzoxathiole Glycine Derivative, S,S-dioxide-N,N'-[3H-2,1-benzoxathiol-3-ylidenebis[[6-hydroxy-2-methyl-5-(1-methylethyl)-3,1-phenylene]methylene]]bis[N-(carboxymethyl)glycine Tetrasodium Salt, N,N'-[(1,1-Dioxido-3H-2,1-benzoxathiol-3-ylidene)bis[[6-hydroxy-2-methyl-5-(1-methylethyl)-3,1-phenylene]methylene]]bis[N-(carboxymethyl)glycine Tetrasodium Salt |
| InChI Formula | InChI=1S/C37H44N2O13S.4Na/c1-19(2)23-11-28(21(5)25(35(23)48)13-38(15-31(40)41)16-32(42)43)37(27-9-7-8-10-30(27)53(50,51)52-37)29-12-24(20(3)4)36(49)26(22(29)6)14-39(17-33(44)45)18-34(46)47;;;;/h7-12,19-20,48-49H,13-18H2,1-6H3,(H,40,41)(H,42,43)(H,44,45)(H,46,47);;;;/q;4*+1/p-4 |
| SMILES | O=C(CN(CC1=C(C)C(C2(C3=CC(C(C)C)=C(C(CN(CC(O[Na])=O)CC(O[Na])=O)=C3C)O)OS(=O)(C4=C2C=CC=C4)=O)=CC(C(C)C)=C1O)CC(O[Na])=O)O[Na] |
| IUPAC Name | sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate |
| Molecular Weight (g/mol) | 844.74 |
| Formula Weight | 844.18 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction