missing translation for 'onlineSavingsMsg'
Learn More

Methylthymol Blue Sodium Salt, TRC

Product Code. 30480029
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30480029

Brand: Toronto Research Chemicals TRCM338655100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylthymol Blue Sodium Salt
CAS 1945-77-3
Molecular Formula C37H40N2Na4O13S
Quantity 100 mg
Synonym 3H-2,1-Benzoxathiole Glycine Derivative, S,S-dioxide-N,N'-[3H-2,1-benzoxathiol-3-ylidenebis[[6-hydroxy-2-methyl-5-(1-methylethyl)-3,1-phenylene]methylene]]bis[N-(carboxymethyl)glycine Tetrasodium Salt, N,N'-[(1,1-Dioxido-3H-2,1-benzoxathiol-3-ylidene)bis[[6-hydroxy-2-methyl-5-(1-methylethyl)-3,1-phenylene]methylene]]bis[N-(carboxymethyl)glycine Tetrasodium Salt
InChI Formula InChI=1S/C37H44N2O13S.4Na/c1-19(2)23-11-28(21(5)25(35(23)48)13-38(15-31(40)41)16-32(42)43)37(27-9-7-8-10-30(27)53(50,51)52-37)29-12-24(20(3)4)36(49)26(22(29)6)14-39(17-33(44)45)18-34(46)47;;;;/h7-12,19-20,48-49H,13-18H2,1-6H3,(H,40,41)(H,42,43)(H,44,45)(H,46,47);;;;/q;4*+1/p-4
SMILES O=C(CN(CC1=C(C)C(C2(C3=CC(C(C)C)=C(C(CN(CC(O[Na])=O)CC(O[Na])=O)=C3C)O)OS(=O)(C4=C2C=CC=C4)=O)=CC(C(C)C)=C1O)CC(O[Na])=O)O[Na]
IUPAC Name sodium 2,2',2'',2'''-((((1,1-dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetate
Molecular Weight (g/mol) 844.74
Formula Weight 844.18
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.