Learn More
Metro™ Double-Deep Top-Track Mobile Unit Kits
Kit provides components for shelving units that fit on tracks for added mobility
Brand:  Metro™ LTTM21C
Additional Details : Weight : 39.62400kg
 View more versions of this product
Product Code. 15917161
  • 1250.00€ / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
Description
Description
  • For use with Metro Super Erecta™ and Super Adjustable™ shelving systems
  • Kit includes posts, casters, caster channels, donut bumpers, roller bearing assemblies and tie-together hardware to connect two units
  • Chrome finish
Order Info
Shelves are sold separately.
Specifications
Specifications
Posts, casters/channel caster assembly, donut bumpers, roller bearing assemblies, tie-together hardware to connect double-deep Top-Track™ two mobile units
33.7 kg
Chrome plated
75 lb.
Metro™ Super Erecta™, Super Adjustable™ shelving system
Chrome
23.25 in.
21 in.
Mobile Unit Kit
59.05 cm
53 cm