Learn More
Metro™ High-Density qwikTRAK™ Track Sets
Product Code. p-4246816
Click to view available options
Depth (English):
30 in.
72 in.
84 in.
96 in.
108 in.
120 in.
132 in.
144 in.
156 in.
168 in.
180 in.
192 in.
204 in.
216 in.
228 in.
240 in.
252 in.
Weight (English):
4 lb.
9 lb.
10 lb.
11 lb.
13 lb.
14 lb.
15 lb.
17 lb.
18 lb.
19 lb.
21 lb.
22 lb.
23 lb.
25 lb.
26 lb.
27 lb.
29 lb.
Unit Size:
Each
Product Code. 15926881
Brand: Metro™ BTS10NA
Only null left
Add to basket
Add to basket
Create tracks up to 21 ft.
  • For use with Metro Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q and MetroMax i shelving systems
  • Sets include tracks and hardware needed to assemble system up to 21 ft. (640cm)
  • 17-823-715 (BTS2.5NA) includes a joining kit to connect the track to other track sets
Specifications
Product Type Track Set
Depth (English) 120 in.
Depth (Metric) 304.8 cm
For Use With (Equipment) Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ MetroMax Q™ and MetroMax i™ shelving system
Includes Tracks and hardware needed to assemble high-density qwikTRAK™ system up to 21 ft.
Weight (English) 14 lb.
Weight (Metric) 6.4 kg