Learn More
Metro™ MetroMax i Glassware Cart, Accessories
For use with MetroMax i carts
Brand:  Metro™ GWBSKT36
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11725659
  • 485.00€ / Each
Estimated Shipment: 17-09-2024
to see stock.
Add to basket
Description
Description
  • 36 in. basket for glassware cart
Specifications
Specifications
Basket
For MetroMax carts
17.8 cm
90 cm
18 in.
35.5 x 18 x 7 in. (90 x 45.7 x 17.8 cm)
7 in.
35.5 in.
Vinyl
45.7 cm