Learn More
Metro™ MetroMax i™ Polymer Lab Worktable, Gray Phenolic Top with 3-Sided Frame
Product Code. 15987041
Click to view available options
Dimensions (L x W x H):
47.75 x 30 x 40.5 in.
Includes:
3 Sided frame
Product Type:
Stationary Table
Unit Size:
Each
Product Code. 15987041
Brand: Metro™ LT48XUPG3
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Keep your lab cleaner with the Metro MetroMax i™ Polymer Lab Worktable, featuring superior corrosion resistance with built-in Microban™ antimicrobial product protection.
These worktables provide the ideal product platform, are modular by design and offer ultimate flexibility based on the application requirements.
  • Advanced polymer construction for corrosive lab environments
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters and accessories must be ordered separately.
Specifications
Product Type Stationary Table
Dimensions (L x W x H) 47.75 x 30 x 40.5 in.
For Use With (Application) Ideal for lab
Height (English) 40.5 in.
Height (Metric) 101.6 cm
Includes 3 Sided frame
Length (English) 47.75 in.
Length (Metric) 121.3 cm
Load Bearing Capacity (English) 600 lb.
Load Bearing Capacity (Metric) 273 kg
Width (English) 30 in.
Width (Metric) 76.2 cm
Material Polymer