Learn More
Metro™ qwikTRAK™ Stationary Intermediate Unit Kits
Product Code. 15916871
Click to view available options
For Use With (Equipment):
Metro MetroMax i and Q Shelving System
Metro Super Erecta, Super Adjustable Shelving System
Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
Material:
Polymer
Stainless Steel
Steel
Weight (English):
15.7 lb.
22.6 lb.
Unit Size:
Each
Product Code. 15916871
Brand: Metro™ BTAK3
Only null left
Add to basket
Add to basket
Includes
  • 1880mm (74 in.) high posts
  • Hardware for connecting one stationary intermediate unit to tracks
  • Hardware for anchoring tracks to the floor
High-density, easy access storage
QwikTRAK storage systems are designed with a track anchored to the floor which allows easy access to all shelves in an efficient storage space.
  • Stationary intermediate unit helps organize storage systems
  • 1880mm (74 in.) high posts
For storage with Super Erecta™, Super Erecta Pro™, Super Adjustable™, MetrMax Q™, or MetroMax i™ shelf storage systems
Order Info
Shelves sold separately
Specifications
Product Type Stationary Intermediate Unit Kit
Finish Metroseal Green Epoxy
For Use With (Equipment) Metro Super Erecta, Super Erecta Pro, Super Adjustable Shelving System
Height (English) 74 in.
Height (Metric) 1880 mm
Includes 74 in. High stationary posts and connecting hardware
Weight (English) 22.6 lb.
Weight (Metric) 10.3 kg
Material Steel