Learn More
Metro™ Stainless Lab Worktable, Gray Phenolic Top and 3-Sided Frame
Click to view available options
Dimensions (L x W x H):
29.75 x 30 x 36 in.
35.75 x 30 x 36 in.
47.75 x 30 x 36 in.
59.75 x 30 x 36 in.
Product Type:
Mobile Table
Stationary Table
Includes:
3-sided Frame
3-sided Frame, Adaptor Insert
Weight (English):
70 lb.
82 lb.
101 lb.
120 lb.
Unit Size:
Each
Description
- Easily cleaned smooth surfaces
- Mobile units can be easily moved to clean behind table area
- Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
| Product Type | Mobile Table |
| Color | Silver |
| Dimensions (L x W x H) | 47.75 x 30 x 36 in. |
| For Use With (Application) | Laboratory |
| Height (English) | 36 in. |
| Height (Metric) | 91.44 cm |
| Includes | 3-sided Frame, Adaptor Insert |
| Length (English) | 47.75 in. |
| Length (Metric) | 121.2 cm |
| Load Bearing Capacity (English) | 600 lb. |
| Show More |