Learn More
Metro™ Gray Label Holders for Wire Shelving
Product Code. 11890900
Click to view available options
Label Size (English):
3 x 1.25 in.
13 x 1.25 in.
19 x 1.25 in.
25 x 1.25 in.
31 x 1.25 in.
43 x 1.25 in.
55 x 1.25 in.
Label Size (Metric):
109.22 x 3.2 cm
139.5 x 3.2 cm
33 x 3.2 cm
48 x 3.2 cm
63.5 x 3.2 cm
7.5 x 3.2 cm
72.5 x 3.2 cm
Unit Size:
Each
Product Code. 11890900
Brand: Metro™ 9990P
Only null left
Add to basket
Edge
Add to basket
Snap-on plastic holders provide easy identification of shelf contents.
  • Gray
  • Hold most commercial 1.25 in. (32 mm) labels
  • Labels not included
Specifications
Product Type Label Holder
Color Gray
For Use With (Equipment) Metro™ Super Erecta™, Super Adjustable™ 2, qwikSLOT™, Drop Mat™ wire shelving, cantilever shelves
Label Size (English) 3 x 1.25 in.
Label Size (Metric) 7.5 x 3.2 cm
Style Solid
Temperature Range (English) -18 to 120.2°F
Temperature Range (Metric) -28 to 49°C
Material Plastic