Learn More
Metro™ MetroMax Q™ Intermediate Shelf
Improve storage in your facility with Metro MetroMax Q shelves.
Brand:  Metro™ MQ2460G
Additional Details : Weight : 10.43000kg
Product Code. 11705649
  • 216.00€ / Each
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
  • Industrial shelving designed for cooler/freezer environments and dry storage
  • Polymer with built-in Microban™ antimicrobial protection inhibits growth of bacteria, mold, mildew, and fungus
Specifications
Specifications
24 in.
Shelf
152.4 cm
60 in.
61 cm