missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I (H-2Kb) Mouse anti-Mouse, PE, Clone: AF6-88.5.5.3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5958-80

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 12727327

  • 71.75€ / Each


In stock, ready to ship: 1
Add to basket

Description

Description

Description: This AF6-88.5.5.3 monoclonal antibody reacts with the H-2Kb MHC class I alloantigen. H-2Kb is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. d, f, j, k, p, q, r, s, u, and v) has not been observed. Applications Reported: This AF6-88.5.5.3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AF6-88.5.5.3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8.
Specifications

Specifications

MHC Class I (H-2Kb)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P01902
H-2K, H-2K(d), H2-K, K-f,H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K(B); H-2K(K); H-2K(Q); MHC H2-K transplantation antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I antigen; MHC class I heavy chain H2-K; class I major histocom
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
AF6-88.5.5.3
PE
H2-K1
Liquid
H2-K1
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14972
Videos
SDS
Documents
Certificates

For Research Use Only.