Learn More
Motic™ MHV-5 Five-Head Observation Optical Bridge, BA210/BA310
Multi-head viewing system suitable for BA210 and BA310 Microscopes. Motic™ MHV-5 Five-Head Observation Optical Bridge, BA210/BA310 facilitates viewing by up to 5 individuals.
Brand: Motic™ 1101001903082
Description
- LED pointer on primary unit.
- Adjustable height support apparatus for secondary unit (2 piece).
- Four additional heads and four pairs of eyepieces required.
Specifications
Five-head Observation Bridge | |
BA210/BA310 Microscopes | |
LED pointer on primary unit. |