Learn More
Motic™ MHV-5 Five-Head Observation Optical Bridge, BA210/BA310
Multi-head viewing system suitable for BA210 and BA310 Microscopes. Motic™ MHV-5 Five-Head Observation Optical Bridge, BA210/BA310 facilitates viewing by up to 5 individuals.
Brand:  Motic™ 1101001903082
Additional Details : Weight : 1.00000kg
Product Code. 12895833
  • 5440.00€ / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
  • LED pointer on primary unit.
  • Adjustable height support apparatus for secondary unit (2 piece).
  • Four additional heads and four pairs of eyepieces required.
Specifications
Specifications
Five-head Observation Bridge
BA210/BA310 Microscopes
LED pointer on primary unit.