Learn More
Robu™ Micro-Immersion-Filter
Robu™ Micro-Immersion-Filter is designed to be suitable for reverse filtration.
Brand:  Robu™ 25104
Additional Details : Weight : 0.04660kg
Product Code. 16863066
  • 110.00€ / Pack of 5
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
Description
Description
  • For reverse filtration
  • Highly resistant to most chemicals such as solvents, acids, and alkalis
Specifications
Specifications
10 to 16 μm
100 mm
6 mm Top, 10 mm Bottom