Learn More
Robu™ Micro-Immersion-Filter
Robu™ Micro-Immersion-Filter is designed to be suitable for reverse filtration.
Brand: Robu™ 25104
Additional Details : Weight : 0.04660kg
Description
- For reverse filtration
- Highly resistant to most chemicals such as solvents, acids, and alkalis
Specifications
10 to 16 μm | |
100 mm | |
6 mm Top, 10 mm Bottom |