Learn More
Kimble Chase™ Micro Separatory Funnels
Kimble Chase™ Micro Separatory Funnels are used in liquid-liquid extractions to separate (partition) the components of a mixture into two immiscible solvent phases of different densities.
Brand:  Kimble Chase™ 298280-0010
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12817120
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Standard taper joints
  • Squibb-type funnel with a PTFE stopcock plug with a 2mm bore
  • Plug is 821001-0002
  • Stopper not included
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and STM E438, Type I, Class A requirements
Specifications
Specifications
Micro Separatory Funnel, Squibb-type
Borosilicate Glass
ST 14/20
10 mL
141 mm