Learn More
SI Analytics™ Micro-Ubbelohde Viscometers
Shop All SI Analytics Products
Click to view available options
Viscosity Range:
4 to 60 cSt
40 to 800 cSt
Unit Size:
Each
Description
- Calibrated, with constant, for manual measurement
- Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, minimum measurement substance and/or rinsing agent quantities, and high-temperature or low temperature measurements
- Delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
- All viscometers are provided with ring marks
- Due to their design, these viscometers are especially suitable for measurement of small liquid quantities and for particularly short running times
- This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
- This constant is determined by multiplication of the constant K with the correction factor F
Specifications
Specifications
| Product Type | Ubbelohde Micro Viscometer |
| Viscosity Range | 40 to 800 cSt |
| Calibrated | Yes |
| Description | 1.26 mm diameter |
| Approximate Constant | 1 |
| Certifications/Compliance | DIN 51562, Part 2 |
| Material | Glass |
| Length (Metric) | 290 mm |
| For Use With (Application) | Manual measurements |
| Volume (Metric) | 3 to 4 mL |