Learn More
Ohaus™ Micro-Well Plate Holder
Product Code. 15752649 Shop All Ohaus Products
Click to view available options
Material:
Plastic
Unit Size:
Pack of 2
Product Code. 15752649
Brand: Ohaus™ 30400233
Only null left
Add to basket
Add to basket
OHAUS™ Micro-Well Plate Holder desigend to securely hold microplates.
  • OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
  • Increase throughput by mixing multiple samples at one time
  • Effortlessly mix up to 52 microtubes or eight microplates
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microplate Holder
Material Plastic
Description Micro-Well Plate Holder, Pack of 2