Learn More
Savillex™ Microcentrifuge Via
Brand:  Savillex™ 20091550
77.68 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 0.80000kg
Product Code. 11987495
  • 220.48€ / Pack of 10
In Stock
to see stock.
Add to basket
Specifications
10,000g
1.5 mL
–200°C to 260°C