Learn More
Ansell MICROCHEM™ 6000
Product Code. 17130975
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
O60A812E
O60A813E
O60A818E
O60A819E
O60T801B
O60T802E
O60T803E
O60T808E
O60T809E
Unit Size:
Each
Product Code. 17130975
Brand: Ansell OR60TA0081807GA1
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Protective suit with multi-layer chemical barrier technology for Type 1 / Level A Gas-tight protection.
  • Excellent chemical protection, >480 mins for 15 chemicals specified by EN 943-2:2002
  • Innovative fabric technology, high performance barrier laminate with scrim reinforcement
  • Lightweight and flexible
  • Highly visible bright orange color for improved worker safety
  • Ultrasonically welded and hot air taped seams
  • DYNAT/YKK Gas-Tight zipper provides protection and performance in the most hostile of chemical environments
  • The zip has an outer zip flap made of the same material as the suit
  • Semi-rigid 3 layer GAG visor provides excellent optical clarity
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases, Tank Cleaning
Material Multi-Layer Non-Woven Barrier Laminate Fabric
Size 3X-Large
Seam Type Welded and Taped
Style O60A818E
Weight (Metric) 230 gsm