Learn More
Ansell Microgard™ 1500 Red Coveralls with Hood
Product Code. p-7074288 Shop All Ansell Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
Pack of 50
Product Code. 15506484
Brand: Ansell RD15S0013804
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Strip, clear up and handle asbestos safely. Ansell Microgard™ 1500 Coveralls with Hood filter particles as small as 3μ
  • Coverall with 3-piece hood
  • Elasticated wrists, waist, and ankles
  • 2-way front zipper with re-sealable storm flap
  • Filter 100% of particles >3 microns
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone-free for spray painting applications
  • Optimised body fit for wearer comfort and safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material SMS Polypropylene
Color Red
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Certifications/Compliance (Europe) EN ISO 13982-1, EN13034
Model RD15-S-00-138
No. of Pockets None
Style Includes Hood