Learn More
3M™ Solus™ 1000 Series Safety Glasses
Scotchgard™ Anti-Fog coating resists fogging for longer than the traditional coatings, even after multiple washes.
Brand: 3M™ 7100078882
This item is not returnable.
View return policy
Description
- Vented, removable foam gasket to help reduce eye exposure
- Padded temples for added comfort
- Removable temple
- Soft nose bridge
- Z87+ Lens marking
- 3M Catalog# S1101SGAF, S1101SGAF-KT, S1102SGAF, S1103SGAF, S1201SGAF, S1201SGAF-KT, S1202SGAF, S1203SGAF
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.