Learn More
Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates
GREENER_CHOICE
Product Code. 10033281
Click to view available options
Binding Property:
High Binding
Medium Binding
Unit Size:
Pack of 100
Product Code. 10033281
Brand: Greiner Bio-One 701070
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use in flexibility in diagnostics. Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates are suitable for transmission measurements.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • For optical measurements
  • Measuring light beam is not deflected by well profile
  • Specifically treated to provide incresed protein binding
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Medium Binding
Format Strip Plate
Array 8 x 12
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
Quantity 100/Cs.