Learn More
Witeg™ Micropipettes
Product Code. 11825844
Click to view available options
Volume (Metric):
20 to 200 μL
200 to 1000 μL
Unit Size:
Each
Product Code. 11825844
Brand: Witeg™ 5 402 901
Only null left
Add to basket
Add to basket
Witeg™ Micropipettes is an accurate, robust, and handy single-channel, microliter pipette.
  • High quality pipette, made in Germany
  • Economically priced
  • Perfect for precise laboratory applications
  • 7 color-coded single-channel models
  • High accuracy and precision according to DIN EN ISO 8655-2
  • Ergonomically designed and low weighted
  • Optimized spring force for comfortable pipetting
  • Robust and stable construction
  • Tip ejector and piston made of stainless steel
  • Suitable for all established brands of pipette tips
  • Completely autoclavable at 121°C (5000mL filter is not autoclavable)
  • Individual certificate and serial number
  • DE-M marked according to German calibration law
Specifications
Accuracy 1.6μL
ClipTip Ejector Tip
Autoclavable Autoclavable
Color Blue
For Use With Precision Dispensing
Number of Channels 1
Type Micropipette
Volume (Metric) 200 to 1000 μL
For Use With (Application) Precision dispensing
Precision ≤ 3μL
Quantity 1 Each